2-Acetylamino-5-bromo-4-methylpyridine 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD18310-5GEA 138 EUR
BLDPBD18310-5G BLDPBD18310-250MG BLDPBD18310-25G BLDPBD18310-0.025G BLDPBD18310-1G
2-Acetylamino-5-bromo-4-methylpyridine 98%
2-Acetylamino-5-bromo-4-methylpyridine

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₉BrN₂O
MW: 229,0700 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06798092
Núm. CAS: 142404-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR