1-Amino-4-bromoisoquinoline 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD182498-5GEA 118 EUR
BLDPBD182498-5G BLDPBD182498-10G BLDPBD182498-250MG BLDPBD182498-0.025 BLDPBD182498-25G BLDPBD182498-1G
1-Amino-4-bromoisoquinoline 97%
1-Amino-4-bromoisoquinoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₇BrN₂
MW: 223,0693 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD05155261
Núm. CAS: 55270-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR