2,6-Diamino-9,10-anthraquinone 98%

BLDPBD17998-100GEA 80 EUR
BLDPBD17998-100G BLDPBD17998-10G BLDPBD17998-25G
2,6-Diamino-9,10-anthraquinone 98%
2,6-Diamino-9,10-anthraquinone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₄H₁₀N₂O₂
MW: 238,2414 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00001234
Núm. CAS: 131-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR