2,2',7,7'-Tetrabromo-9,9'-spirobifluorene 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD17926-1GEA 9 EUR
BLDPBD17926-1G BLDPBD17926-25G BLDPBD17926-5G BLDPBD17926-10G BLDPBD17926-100G
2,2',7,7'-Tetrabromo-9,9'-spirobifluorene 98%
2,2',7,7'-Tetrabromo-9,9'-spirobifluorene

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₅H₁₂Br₄
MW: 631,9800 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08704220
Núm. CAS: 128055-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR