(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate 98%

BLDPBD1791-250MGEA 6 EUR
BLDPBD1791-250MG BLDPBD1791-100G BLDPBD1791-10G BLDPBD1791-25G BLDPBD1791-500G BLDPBD1791-5G BLDPBD1791-1G
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate 98%
(2R,3S,4R,5R)-5-((Benzoyloxy)methyl)-3-fluorotetrahydrofuran-2,4-diyl dibenzoate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₂₆H₂₁FO₇
MW: 464,44 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 97614-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR