Methyl 6-cyanonicotinate 97%

BLDPBD174329-100MGEA 18 EUR
BLDPBD174329-100MG BLDPBD174329-250MG BLDPBD174329-1G
Methyl 6-cyanonicotinate 97%
Methyl 6-cyanonicotinate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₆N₂O₂
MW: 162,145 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09927471
Núm. CAS: 89809-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR