tert-Butyl 5-formylindoline-1-carboxylate 98%

BLDPBD173930-1GEA 168 EUR
BLDPBD173930-1G BLDPBD173930-250MG BLDPBD173930-5G BLDPBD173930-100MG
tert-Butyl 5-formylindoline-1-carboxylate 98%
tert-Butyl 5-formylindoline-1-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₁₇NO₃
MW: 247,29 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD12408236
Núm. CAS: 879887-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR