1,1,2-Trimethyl-1H-benz[e]indole 98%

BLDPBD17084-10GEA 13 EUR
BLDPBD17084-10G BLDPBD17084-0.025G BLDPBD17084-25G BLDPBD17084-500G BLDPBD17084-100G BLDPBD17084-5G
1,1,2-Trimethyl-1H-benz[e]indole 98%
1,1,2-Trimethyl-1H-benz[e]indole

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₅H₁₅N
MW: 209,29 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00082627
Núm. CAS: 41532-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR