Methyl-2-benzoylbenzoate 98%

BLDPBD17079-25GEA 7 EUR
BLDPBD17079-25G BLDPBD17079-100G BLDPBD17079-500G BLDPBD17079-0.5G BLDPBD17079-1G BLDPBD17079-1000G
Methyl-2-benzoylbenzoate 98%
Methyl-2-benzoylbenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₅H₁₂O₃
MW: 240,2540 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00017187
Núm. CAS: 606-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR