Methyl-3-iodo-1H-indazole-4-carboxylate 95+%
|
|
BLDPBD168073-250MGEA
29
EUR
BLDPBD168073-250MG
BLDPBD168073-5G
BLDPBD168073-100MG
BLDPBD168073-1G
Methyl-3-iodo-1H-indazole-4-carboxylate 95+%
Methyl-3-iodo-1H-indazole-4-carboxylate
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₉H₇IN₂O₂ MW: 302,0686 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD07781588 Núm. CAS: 885521-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...