2-Bromo-6-methoxyaniline 97%

BLDPBD167982-5GEA 142 EUR
BLDPBD167982-5G BLDPBD167982-250MG BLDPBD167982-10G BLDPBD167982-1G BLDPBD167982-25G
2-Bromo-6-methoxyaniline 97%
2-Bromo-6-methoxyaniline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₈BrNO
MW: 202,0500 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08741448
Núm. CAS: 5473-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR