1H-Indazole-4-boronic acid 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD167747-1GEA 50 EUR
BLDPBD167747-1G BLDPBD167747-25G BLDPBD167747-5G BLDPBD167747-250MG
1H-Indazole-4-boronic acid 97%
1H-Indazole-4-boronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₇H₇BN₂O₂
MW: 161,9500 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD09878901
Núm. CAS: 1023595-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR