6-Bromo-2-chloro-3-ethylquinoline 98+%

BLDPBD167658-250MGEA 82 EUR
BLDPBD167658-250MG BLDPBD167658-100MG BLDPBD167658-1G
6-Bromo-2-chloro-3-ethylquinoline 98+%
6-Bromo-2-chloro-3-ethylquinoline

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₉BrClN
MW: 270,553 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09029946
Núm. CAS: 409346-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR