(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid 97%

BLDPBD167180-10GEA 396 EUR
BLDPBD167180-10G BLDPBD167180-25G BLDPBD167180-5G BLDPBD167180-250MG BLDPBD167180-1G
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid 97%
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₇NO₅
MW: 243,26 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01861764
Núm. CAS: 198646-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR