4-Cyano-4'-propylbiphenyl 98%

BLDPBD16328-1GEA 6 EUR
BLDPBD16328-1G BLDPBD16328-25G BLDPBD16328-5G BLDPBD16328-100G
4-Cyano-4'-propylbiphenyl 98%
4-Cyano-4'-propylbiphenyl

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₆H₁₅N
MW: 221,2970 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00505987
Núm. CAS: 58743-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR