α-Bromo-5-chloro-2-nitrotoluene 98%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD162876-1GEA
18
EUR
BLDPBD162876-1G
BLDPBD162876-10G
BLDPBD162876-25G
BLDPBD162876-5G
α-Bromo-5-chloro-2-nitrotoluene 98%
α-Bromo-5-chloro-2-nitrotoluene
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₅BrClNO₂ MW: 250,4771 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD11848697 Núm. CAS: 31577-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...