5-Hydroxyisophthalonitrile 97%

BLDPBD162582-250MGEA 11 EUR
BLDPBD162582-250MG BLDPBD162582-5G BLDPBD162582-1G BLDPBD162582-10G BLDPBD162582-25G
5-Hydroxyisophthalonitrile 97%
5-Hydroxyisophthalonitrile

Storage condition: Inert atmosphere, Room Temperature

Formula: C8H4N2O
MW: 144,1302 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11865326
Núm. CAS: 79370-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR