2-Chloro-3-ethoxy-6-fluorophenylboronic acid 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD161613-250MGEA 17 EUR
BLDPBD161613-250MG BLDPBD161613-1G BLDPBD161613-5G
2-Chloro-3-ethoxy-6-fluorophenylboronic acid 97%
2-Chloro-3-ethoxy-6-fluorophenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₉BClFO₃
MW: 218,4200 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09800880
Núm. CAS: 957120-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR