Methyl-4-bromo-3-(trifluoromethyl)benzoate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD161385-250MGEA 6 EUR
BLDPBD161385-250MG BLDPBD161385-5G BLDPBD161385-1G BLDPBD161385-25G BLDPBD161385-10G
Methyl-4-bromo-3-(trifluoromethyl)benzoate 98%
Methyl-4-bromo-3-(trifluoromethyl)benzoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₉H₆BrF₃O₂
MW: 283,0420 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD10566409
Núm. CAS: 107317-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR