Ethyl-4-amino-3-fluorobenzoate 97%

BLDPBD161349-5GEA 41 EUR
BLDPBD161349-5G BLDPBD161349-1G BLDPBD161349-250MG BLDPBD161349-100MG BLDPBD161349-25G BLDPBD161349-10G
Ethyl-4-amino-3-fluorobenzoate 97%
Ethyl-4-amino-3-fluorobenzoate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₁₀FNO₂
MW: 183,1796 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD07782043
Núm. CAS: 73792-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR