1-Bromo-2,3-difluoro-4-nitrobenzene 98%

BLDPBD161308-25GEA 1098 EUR
BLDPBD161308-25G BLDPBD161308-5G BLDPBD161308-10G BLDPBD161308-1G BLDPBD161308-250MG
1-Bromo-2,3-difluoro-4-nitrobenzene 98%
1-Bromo-2,3-difluoro-4-nitrobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₂BrF₂NO₂
MW: 237,9900 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09839215
Núm. CAS: 1003708-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR