2-Fluoro-m-anisidine 98%
|
|
BLDPBD161264-250MGEA
31
EUR
BLDPBD161264-250MG
BLDPBD161264-5G
BLDPBD161264-1G
BLDPBD161264-10G
BLDPBD161264-100MG
BLDPBD161264-25G
2-Fluoro-m-anisidine 98%
2-Fluoro-m-anisidine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₇H₈FNO MW: 141,1400 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD11042216 Núm. CAS: 801282-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...