2-Bromo-3,4-difluoroaniline 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD160832-1GEA 9 EUR
BLDPBD160832-1G BLDPBD160832-250MG BLDPBD160832-5G BLDPBD160832-10G BLDPBD160832-100G BLDPBD160832-25G
2-Bromo-3,4-difluoroaniline 97%
2-Bromo-3,4-difluoroaniline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₄BrF₂N
MW: 208,00 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD11035924
Núm. CAS: 1092349-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR