Methyl-3,3-dimethoxypropionate 97%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD159839-10GEA
13
EUR
BLDPBD159839-10G
BLDPBD159839-100G
BLDPBD159839-25G
BLDPBD159839-500G
BLDPBD159839-5G
Methyl-3,3-dimethoxypropionate 97%
Methyl-3,3-dimethoxypropionate
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₆H₁₂O₄ MW: 148,16 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00010650 Núm. CAS: 7424-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...