6,7-Dimethoxy-3,4-dihydroisoquinoline 98%
|
|
BLDPBD159185-5GEA
55
EUR
BLDPBD159185-5G
BLDPBD159185-1G
6,7-Dimethoxy-3,4-dihydroisoquinoline 98%
6,7-Dimethoxy-3,4-dihydroisoquinoline
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₁H₁₃NO₂ MW: 191,23 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00266748 Núm. CAS: 3382-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...