4-Chloroisoquinoline 95%

BLDPBD159072-1GEA 49 EUR
BLDPBD159072-1G BLDPBD159072-250MG BLDPBD159072-5G BLDPBD159072-25G
4-Chloroisoquinoline 95%
4-Chloroisoquinoline

Storage condition: Sealed in dry, 2-8°C

Formula: C₉H₆ClN
MW: 163,6036 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00234490
Núm. CAS: 1532-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR