Ethyl-4-hydroxyquinoline-2-carboxylate 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD158985-1GEA 42 EUR
BLDPBD158985-1G BLDPBD158985-250MG BLDPBD158985-5G
Ethyl-4-hydroxyquinoline-2-carboxylate 95%
Ethyl-4-hydroxyquinoline-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₁NO₃
MW: 217,2200 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD01863732
Núm. CAS: 24782-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR