4-Cyano-4'-ethoxybiphenyl 97%
|
|
BLDPBD15894-250MGEA
7
EUR
BLDPBD15894-250MG
BLDPBD15894-1G
BLDPBD15894-5G
4-Cyano-4'-ethoxybiphenyl 97%
4-Cyano-4'-ethoxybiphenyl
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₅H₁₃NO MW: 223,2700 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01218033 Núm. CAS: 58743-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...