tert-Butyl [2-(4-aminophenoxy)ethyl]carbamate 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD158704-1GEA 130 EUR
BLDPBD158704-1G BLDPBD158704-250MG BLDPBD158704-5G
tert-Butyl [2-(4-aminophenoxy)ethyl]carbamate 97%
tert-Butyl [2-(4-aminophenoxy)ethyl]carbamate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₃H₂₀N₂O₃
MW: 252,309 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD11922406
Núm. CAS: 159184-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR