2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD158557-250MGEA 62 EUR
BLDPBD158557-250MG BLDPBD158557-100MG
2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride 97%
2-Methyl-1,3,7-triazaspiro[4.5]dec-1-en-4-one hydrochloride

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₃N₃O·ClH
MW: 203,669 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11869758
Núm. CAS: 1214028-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR