3-Bromo-1H-pyrazolo[4,3-b]pyridine 98%
|
|
BLDPBD158457-25GEA
666
EUR
BLDPBD158457-25G
BLDPBD158457-5G
BLDPBD158457-10G
BLDPBD158457-250MG
BLDPBD158457-1G
3-Bromo-1H-pyrazolo[4,3-b]pyridine 98%
3-Bromo-1H-pyrazolo[4,3-b]pyridine
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₆H₄BrN₃ MW: 198,0201 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD11518974 Núm. CAS: 633328-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...