1-(2,3-Dihydro-7-benzofuranyl)ethanone 95%

BLDPBD158218-1GEA 120 EUR
BLDPBD158218-1G BLDPBD158218-5G BLDPBD158218-100MG BLDPBD158218-250MG
1-(2,3-Dihydro-7-benzofuranyl)ethanone 95%
1-(2,3-Dihydro-7-benzofuranyl)ethanone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₀O₂
MW: 162,1852 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11656624
Núm. CAS: 170730-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR