1,7-Naphthyridin-2(1H)-one 95%

BLDPBD157839-100MGEA 129 EUR
BLDPBD157839-100MG BLDPBD157839-1G BLDPBD157839-250MG
1,7-Naphthyridin-2(1H)-one 95%
1,7-Naphthyridin-2(1H)-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆N₂O
MW: 146,14 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08236271
Núm. CAS: 54920-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR