2,4'-Dichloro-2'-o-chlorobenzoylacetanilide 95+%

Proveedor: BLD PHARMATECH GMBH

BLDPBD15756-5GEA 128 EUR
BLDPBD15756-5G BLDPBD15756-1G BLDPBD15756-250MG
2,4'-Dichloro-2'-o-chlorobenzoylacetanilide 95+%
2,4'-Dichloro-2'-o-chlorobenzoylacetanilide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₅H₁₀Cl₃NO₂
MW: 342,6044 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00000927
Núm. CAS: 14405-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR