Z-8-dodecen-1-ol 90%

BLDPBD156781-100MGEA 79 EUR
BLDPBD156781-100MG BLDPBD156781-250MG BLDPBD156781-1G
Z-8-dodecen-1-ol 90%
Z-8-dodecen-1-ol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₂H₂₄O
MW: 184,31 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00065431
Núm. CAS: 40642-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR