(Z)-Octadec-13-en-1-yl acetate 90%

BLDPBD156760-1GEA 170 EUR
BLDPBD156760-1G BLDPBD156760-100MG BLDPBD156760-250MG
(Z)-Octadec-13-en-1-yl acetate 90%
(Z)-Octadec-13-en-1-yl acetate

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₀H₃₈O₂
MW: 310,5145 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 60037-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR