4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane 97%

BLDPBD15641-25GEA 235 EUR
BLDPBD15641-25G BLDPBD15641-250MG BLDPBD15641-100MG BLDPBD15641-1G BLDPBD15641-10G BLDPBD15641-5G BLDPBD15641-100G
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₄H₂₃BO₄
MW: 266,1410 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD04115656
Núm. CAS: 680596-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR