trans-2-Hexenyl acetate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD154682-25GEA 13 EUR
BLDPBD154682-25G
trans-2-Hexenyl acetate 98%
trans-2-Hexenyl acetate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₁₄O₂
MW: 142,20 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00009474
Núm. CAS: 2497-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR