6-Aminocyclohex-3-enecarboxylic acid hydrochloride 98%

BLDPBD154647-1GEA 416 EUR
BLDPBD154647-1G BLDPBD154647-250MG BLDPBD154647-100MG
6-Aminocyclohex-3-enecarboxylic acid hydrochloride 98%
6-Aminocyclohex-3-enecarboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₁₂ClNO₂
MW: 177,62 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00145419
Núm. CAS: 97945-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR