trans-1-Ethoxy-4-(2-(4-(4-ethylcyclohexyl)phenyl)ethynyl)benzene 95%

BLDPBD154604-100GEA 130 EUR
BLDPBD154604-100G BLDPBD154604-25G BLDPBD154604-5G
trans-1-Ethoxy-4-(2-(4-(4-ethylcyclohexyl)phenyl)ethynyl)benzene 95%
trans-1-Ethoxy-4-(2-(4-(4-ethylcyclohexyl)phenyl)ethynyl)benzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₄H₂₈O
MW: 332,4785 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11053409
Núm. CAS: 334826-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR