1-(Naphthalen-1-yl)piperazine 97%

BLDPBD15439-250MGEA 116 EUR
BLDPBD15439-250MG BLDPBD15439-1G BLDPBD15439-100MG
1-(Naphthalen-1-yl)piperazine 97%
1-(Naphthalen-1-yl)piperazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₄H₁₆N₂
MW: 212,29 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00600021
Núm. CAS: 57536-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR