Tiofeno-2-carbaldehído-oxima 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD153954-5GEA 32 EUR
BLDPBD153954-5G BLDPBD153954-25G
Tiofeno-2-carbaldehído-oxima 98%
Tiofeno-2-carbaldehído-oxima

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₅H₅NOS
MW: 127,17 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00022509
Núm. CAS: 29683-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR