Tetraphenylphosphonium tetraphenylborate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD153692-100GEA 342 EUR
BLDPBD153692-100G BLDPBD153692-1G BLDPBD153692-25G BLDPBD153692-5G
Tetraphenylphosphonium tetraphenylborate 98%
Tetraphenylphosphonium tetraphenylborate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄₈H₄₀BP
MW: 658,6160 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00145389
Núm. CAS: 15525-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR