Tetraethylammonium toluene-4-sulphonate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD153368-10GEA 11 EUR
BLDPBD153368-10G BLDPBD153368-500G BLDPBD153368-5G BLDPBD153368-100G BLDPBD153368-25G
Tetraethylammonium toluene-4-sulphonate 98%
Tetraethylammonium toluene-4-sulphonate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₅H₂₇NO₃S
MW: 301,4500 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00011831
Núm. CAS: 733-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR