2-((tert-Butoxycarbonyl)amino)-2-(p-tolyl)acetic acid 95%

BLDPBD152690-100MGEA 49 EUR
BLDPBD152690-100MG
2-((tert-Butoxycarbonyl)amino)-2-(p-tolyl)acetic acid 95%
2-((tert-Butoxycarbonyl)amino)-2-(p-tolyl)acetic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₁₉NO₄
MW: 265,3050 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03789598
Núm. CAS: 28044-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR