9-Ethyl-3-nitro-9H-carbazole 98%

BLDPBD15268-5GEA 82 EUR
BLDPBD15268-5G BLDPBD15268-1G
9-Ethyl-3-nitro-9H-carbazole 98%
9-Ethyl-3-nitro-9H-carbazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₄H₁₂N₂O₂
MW: 240,2573 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00022215
Núm. CAS: 86-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR