Ethyl arachidate 95%
|
|
BLDPBD1526-5GEA
182
EUR
BLDPBD1526-5G
BLDPBD1526-1G
BLDPBD1526-25G
Ethyl arachidate 95%
Ethyl arachidate
Storage condition: Sealed in dry, Store in freezer, under -20°C
Formula:
C₂₂H₄₄O₂ MW: 340,5800 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00056233 Núm. CAS: 18281-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...