sulphacarbamide 98%

BLDPBD152173-100MGEA 63 EUR
BLDPBD152173-100MG
sulphacarbamide 98%
Sulphacarbamide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₉N₃O₃S
MW: 215,2297 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00025437
Núm. CAS: 547-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR