Quinizarin green 95%
|
|
BLDPBD151710-5GEA
7
EUR
BLDPBD151710-5G
BLDPBD151710-10G
Quinizarin green 95%
Quinizarin green
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₂₈H₂₂N₂O₂ MW: 418,496 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00001195 Núm. CAS: 128-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...