Sodium (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoate 99%

BLDPBD151503-25GEA 6 EUR
BLDPBD151503-25G BLDPBD151503-100G
Sodium (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoate 99%
Sodium (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₂₁NaO₁₂
MW: 380,27 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD06658158
Núm. CAS: 27297-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR